Learn More
Diagnostic Biosystems PERMAYELLOW/HRP PermaYellow/HRP
Supplier:  Diagnostic Biosystems K060
Encompass_Preferred
PermaYellow/HRP is a substrate chromogen system for use with horseradish peroxidase (HRP) detection in immunohistochemistry (IHC) or in situ hybridizations (ISH). PermaYellow/HRP produces a distinct bright yellow that can easily be distinguished from other stains.
Catalog No. NC2049149
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.