Learn More
Invitrogen™ eBioscience™ Permeabilization Buffer (10X)
Catalog No. 501129059
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
50-112-9059 100 mL
1 options
Contains Sodium azide
Catalog No. 50-112-9059 Supplier Invitrogen™ Supplier No. 00833356
  • $146.00
    $109.65 / Each of 1
    Save $36.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart
Contains Sodium azide
Can be used to fix and permeabilize cells prior to performing intracellular staining of cytokines and other cytoplasmic and/or nuclear antigens.
Used in conjunction with the Intracellular (IC) Fixation Buffer (cat. 00-8222) or Foxp3/Transcription Factor Fixation/Permeabilization Concentrate and Diluent (cat. 00-5521). These solutions have been specially formulated for reducing non-specific staining of fluorochrome-labeled antibodies and increasing fluorescence signal to noise ratios.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 mL
Reagent Type Permeabilization Reagent
Product Line eBioscience
Product Type Permeabilization Buffer
For Research Use Only. Not for use in diagnostic procedures.