Learn More
Bioassay Systems QuantiChrom™ Peroxidase Assay Kit. For quantitative determination of peroxidase enzyme activity.
SDP
Catalog No. 502255858 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of peroxidase enzyme activity. Key Features: Fast and sensitive. Use as little as 10 µL samples. Linear detection range: colorimetric assays 2 to 50 U/L, fluorimetric assays 0.1 to 5 U/L peroxidase. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD570nm, or FL530/585nm. Samples: Biological. Species: All. Procedure: Assay takes 20 min. Kit size: 100 tests. Detection limit: OD, FL: 4, 0.8 U/L.
PEROXIDASES (EC number 1.11.1.x) catalyze the following oxidation-reduction reactions:
ROOR + electron donor (2 e-) + 2H+ → ROH + R'OH.
This item is not returnable. View return policy
Catalog No. 50-225-5858 Supplier Bioassay Systems Supplier No. D2PD100
For quantitative determination of peroxidase enzyme activity. Key Features: Fast and sensitive. Use as little as 10 µL samples. Linear detection range: colorimetric assays 2 to 50 U/L, fluorimetric assays 0.1 to 5 U/L peroxidase. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD570nm, or FL530/585nm. Samples: Biological. Species: All. Procedure: Assay takes 20 min. Kit size: 100 tests. Detection limit: OD, FL: 4, 0.8 U/L.
PEROXIDASES (EC number 1.11.1.x) catalyze the following oxidation-reduction reactions:
ROOR + electron donor (2 e-) + 2H+ → ROH + R'OH.