Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Peroxidase (Horseradish), EIA Grade

Catalog No. 89956435 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-956-435 5000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-956-435 Supplier Abnova™ Supplier No. P5313
Add to Cart
Add to Cart

Used for Enzyme

#REF!

Specifications

For Use With (Application) Enzyme Activity
Molecular Weight (g/mol) 40kDa
Name Peroxidase (Horseradish), EIA Grade
Preparation Method Native protein chromatographically purified from Horseradish Roots
Quantity 5000 U
Storage Requirements Store at 4°C. For long term storage store at -20°C.
Regulatory Status RUO
Common Name PEROXIDASE
Assays Immunoconjugation
Biological Activity ≥500 units per mg protein. One Unit decomposes 1 umole of H2O2 per minute at 25°C, pH 7.0 using aminoantipyrine and phenol.
Species Plants
Protein Tag None
Expression System Native protein chromatographically purified from Horseradish Roots
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.