Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ PeroxyGlow A
SDP

Catalog No. 467509601
Change view
Click to view available options
Quantity:
6 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-750-9601 6 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 46-750-9601 Supplier R&D Systems™ Supplier No. 467509601

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

R&D Systems™ PeroxyGlow™ A is a chemiluminescent substrate for horseradish peroxidase (HRP) generating light that can be quantified in a suitable plate reader for luminescence.

  • Component of the Universal Chemiluminescent Assay Kits
  • Includes Dimethyl sulfoxide (CAS No) 67-68-5

Specifications

Content And Storage Store the unopened product at 2 to 8°C
Quantity 6 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.