Learn More
Abcam Peroxynitrite Assay Kit, Fluorometric, Cell-Based, Quantitative, 100 Tests
Description
- Assay type: Cell-based
- Detection method: Fluorescent
- Results type: Quantitative
- Usage: Suitable for use with a microplate reader
- Cited in: > 5 publications
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Peroxynitrite |
| For Use With (Equipment) | Microplate Reader |
| Detection Location | In-Solution Detection |
| Format | Kit |
| No. of Reactions | 100 Tests |
| Detection Method | Fluorescence |
| Sample Type | Cells |
| Quantity | 100 Tests |
| For Use With (Application) | Peroxynitrite measurement |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.