Learn More
Sterlitech Corporation Polyethersulfone (PES) Membrane Filters, 0.45 Micron, 90mm, 25/Pk
SDP
Supplier:  Sterlitech Corporation PES459025
Encompass_Preferred
Sterlitech PES (polyethersulfone) membrane filters, 0.45 micron pore size, 90 mm diameter, pack of 25. Hydrophilic, low protein/drug binding, minimal extractables, and broad chemical tolerance; ideal for pharmaceutical, medical, and life science applications.
Catalog No. NC9978527
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.