Learn More
Sterlitech Corporation PES Syringe Filters, Sterlitech, 0.22 Micron, 13mm, 100/Pk
SDP
Supplier:  Sterlitech Corporation 1470353
Encompass_Preferred
PES Syringe Filter Applications:* Sterilize biological fluids, serum or tissue culture media additives* Sample preparation of aqueous solutions* High throughput, low binding filter* Units for sterile aqueous filtrations* Filter probe and hybridization solutions to reduce backgrounds* Sample preparation of protein-based* HPLC solutions* High throughputs when sterilizing or clarifying even the most viscous proteinaceous solutions
Catalog No. 50-195-2312
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.