Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Bentranil, MilliporeSigma™ Supelco™

Catalog No. 111012307 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2307 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2307 Supplier MilliporeSigma Supelco Supplier No. 3787225MG

Bentranil is a herbicide, which is used for the postemergence control of annual weeds in cereal crops, rice and maize.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 1022-46-4
Physical Form Neat
Linear Formula C14H9 NO2
MDL Number MFCD00043598
Melting Point 123°C to 125°C
Boiling Point 189°C to 192°C (8 mmHg)
Synonym 2-Phenyl-3,1-benzoxazin-4-one
Type Analytical Standard
Quantity 25 mg
Formula Weight 223.23
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.