Learn More
Description
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 103-17-3 |
| Physical Form | Neat |
| Linear Formula | C13H10 Cl2 S |
| MDL Number | MFCD00055267 |
| Synonym | 4-Chlorobenzyl 4-chlorophenyl sulfide |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 269.19 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.