Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Dimoxystrobin Metabolite 505M09, Analytical Standard, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111004914 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-4914 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-4914 Supplier MilliporeSigma Supelco Supplier No. 9312710MG

Dimoxystrobin metabolite 505M09 is a type of pesticide metabolite which belongs to strobilurin class of fungicides.

TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
Linear Formula C19H20N2O5
Synonym (E)-2-[(5-Hydroxycarbonyl-2-methylphenoxy)methyl]-alpha-methoxyimino-N-methylphenylacetamide; (E)-o-[(5-Hydroxycarbonyl-2-methylphenoxy)methyl]-2-methoxyimino-N-methylphenylacetamide
Quantity 10 mg
Formula Weight 356.37
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.