Learn More
PESTANAL™ Ditalimfos Solution, 100 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 5131-24-8 |
| Linear Formula | C12H14 NO4 PS |
| MDL Number | MFCD00055502 |
| Type | Analytical Standard |
| Quantity | 2 mL |
| Formula Weight | 299.28 |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 100 μg/mL in acetonitrile |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.