Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Ethanethiol (ethyl mercaptan), Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111020614 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0614 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0614 Supplier MilliporeSigma Supelco Supplier No. 506818

missing translation for 'validMainframeMsg'

Specifications

Packaging Ampule
CAS 75-08-1
Product Line PESTANAL
Vapor Pressure 8.51 psi (20°C)
Vapor Density 2.1 (vs. air)
Density 0.839 g/mL (at 25°C)
Physical Form Neat
Linear Formula C2H5SH
MDL Number MFCD00004887
Boiling Point 35°C
Synonym Ethanethiol; Ethyl mercaptan; Mercaptan C2
Quantity 1000 mg
Formula Weight 62.13
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.