Learn More
PESTANAL™ KDWR Pesticides Kit 1,000 μg/mL each component in Methanol, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| Packaging | Ampule |
| Product Line | PESTANAL |
| CAS | 67-56-1 |
| Flash Point | 9°C |
| Type | Analytical Standard |
| Quantity | 5 x 1 mL |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 1,000 μg/mL each component in methanol |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.