Learn More
PESTANAL™ Pesticide Standard 17 Solution, Analytical Standard, According to DIN 38407, in Acetonitrile, MilliporeSigma™ Supelco™
Catalog No. 111017379 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mL
Catalog No. Quantity
11-101-7379 2 mL
1 options
Catalog No. 11-101-7379 Supplier MilliporeSigma Supelco Supplier No. 369782ML
Specifications
Description 10 μg/mL each: atrazin, atrazin-desethyl, cyanazin, sebuthylazin, simazin, terbuthylazin, hexazinon, chlortoluron, diuron, isoproturon, linuron, methabenzthiazuron, metobromuron, metoxuron, monolinuron, metazoachlor, metolachlor
Product Line PESTANAL
Quantity 2 mL
Grade Analytical Standard
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH