Learn More
PESTANAL™ Tiadinil, ≥99.0% (HPLC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| Packaging | Bottomless Glass Bottle with Inserted Fused Cone |
| Product Line | PESTANAL |
| CAS | 223580-51-6 |
| Physical Form | Neat |
| Linear Formula | C11H10 ClN3 OS |
| Melting Point | 111°C to 113°C |
| Synonym | 5-(3-Chloro-4-methylanilinocarbonyl)-4-methyl-1,2,3-thiadiazole; N-(3-Chloro-4-methylphenyl)-4-methyl-1,2,3-thiadiazole-5-carboxamide |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 267.73 |
| Show More |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.