Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Vamidothion Solution, 100 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Catalog No. 111019722 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9722 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9722 Supplier MilliporeSigma Supelco Supplier No. 329312ML

Vamidothion belongs to the class of organophosphorus pesticides (OPs) which act as cholinesterase inhibitors and are utilized for protection of crops against spiders, mites, aphids, beetles, caterpillars, etc.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 2275-23-2
Flash Point 2°C - Closed Cup
Linear Formula C8H18 NO4 PS2
MDL Number MFCD00055442
Synonym O,O-Dimethyl S-[2-(1-methyl-2-methylamino-2-oxoethylthio)ethyl] phosphorothioate
Type Analytical Standard
Quantity 2 mL
Formula Weight 287.34
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 100 μg/mL in acetonitrile
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.