Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pesticide Degradation Check Solution, various concentrations in isooctane, Ultra Scientific

Catalog No. USISM4501
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USISM4501 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USISM4501 Supplier Agilent Technologies Supplier No. ISM4501
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Pesticide Degradation Check Solution
CAS 540-84-1, 72-20-8, 50-29-3
CAS Max % 99.99, 0.0, 0.0
Analyte or Component Names Isooctane,endrin,4
Element Isooctane
Solvent or Matrix Isooctane
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) Various concentrations
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.