Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pesticide Kit, SPEX CertiPrep™
SDP

Catalog No. SPXPRKIT Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
10 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SPXPRKIT 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPXPRKIT Supplier SPEX CertiPrep Supplier No. SPXPRKIT

missing translation for 'validMainframeMsg'

Use these Organic Certified Reference Materials for a wide range of HPLC/GC/LC applications. SPEX CertiPrep™ Pesticide Kits containing 10-multi-compound mixes are ideal for pesticide screening, food safety and QuEChERS applications.

  • Pesticide Kit containing 10-multi-compound mixes
  • A total of 144 compounds in acetronitrile
  • Available and sold in 1mL ampules that are supplied with a pre-labeled transfer/storage vial

Warranty and Services

1 year

Specifications

Chemical Name or Material Trifloxystrobin
Boscalid
Fenoxycarb
Piperonyl butoxide
Tebufenpyrad
Iprodione
Imid
Packaging Glass Ampule
Assay Percent Range 99.5
99.5
99
95.8
99.5
99.5
99.5
99.5
99.5
99.5
99.5
99.5
99.5
97.6
99.2
99.3
99.5
98.5
99.4
98.8
99.4
99.5
99.2
99.5
98.3
99.5
99.4
99.3
99.5
99.5
99.5
97.4
99.9
98.6
98.4
98.9
99.5
99.6
99.5
99.5
98.5
99
96
98.9
99
99.4
99.1
98.8
97.5
99.5
98
99
99.6
99.5
99.5
99.2
99.2
99.5
98
99.5
98
98.2
99.5
99.5
99.9
99.9
99.5
98.6
99.2
99.5
99.5
99.5
99.5
98.4
98.8
99.1
99.5
95
99.7
99.2
99.4
99.5
99.5
98.4
99.4
97.5
99.5
98.9
99
99.5
98.4
99
98
98.2
97.7
98.1
99.4
25
99.3
99
99.5
99.5
99.5
98
97.5
99.5
99
98.7
99.5
99.6
99.4
98.6
99.5
97
97
99.5
99.5
96.7
94.2
97
99.5
98.1
98
98.4
98
99.5
99.5
98.6
99.4
98.4
99
99.7
99
99.5
99
99
98
97.7
99
99
98
89.4
99.5
99
Concentration 100 μg/mL
Quantity 10 x 1 mL
Formula Weight C20H19F3N2O4
C18H12Cl2N2O
C17H19NO4
C19H30O5
C18H24ClN
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.