Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pesticide Kit #3, Restek™
SDP

Catalog No. 06720003 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-720-003 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-720-003 Supplier Restek Corporation Supplier No. 32404

  • Aroclor and toxaphene standards may exhibit lot-to-lot variation.

  • Calibration mixes only for CLP 04.1. Includes pesticide standard mixes A & B at 16x CRQL with surrogates


Compounds:

32003: Pesticide Standard Mix A w/Surrogates

α-BHC (319-84-6), 8 μg/mL
γ-BHC (Lindane) (58-89-9), 8 μg/mL
4,4'-DDD (72-54-8), 16 μg/mL
4,4'-DDT (50-29-3), 16 μg/mL
Decachlorobiphenyl (SS) (2051-24-3), 16 μg/mL
Dieldrin (60-57-1), 16 μg/mL
Endosulfan I (959-98-8), 8 μg/mL
Endrin (72-20-8), 16 μg/mL
Heptachlor (76-44-8), 8 μg/mL
Methoxychlor (72-43-5), 80 μg/mL
2,4,5,6-Tetrachloro-m-xylene (SS) (877-09-8), 8 μg/mL

32004: Pesticide Standard Mix B w/Surrogates
Aldrin (309-00-2), 8 μg/mL
β-BHC (319-85-7), 8 μg/mL
δ-BHC (319-86-8), 8 μg/mL
cis-Chlordane (5103-71-9), 8 μg/mL
trans-Chlordane (5103-74-2), 8 μg/mL
4,4'-DDE (72-55-9), 16 μg/mL
Decachlorobiphenyl (SS) (2051-24-3), 16 μg/mL
Endosulfan II (33213-65-9), 16 μg/mL
Endosulfan sulfate (1031-07-8), 16 μg/mL
Endrin aldehyde (7421-93-4), 16 μg/mL
Endrin ketone (53494-70-5), 16 μg/mL
Heptachlor epoxide (isomer B) (1024-57-3), 8 μg/mL
2,4,5,6-Tetrachloro-m-xylene (SS) (877-09-8), 8 μg/mL

32005: Toxaphene Solutions
Toxaphene (8001-35-2)

32007: Aroclor 1221
Aroclor 1221

32008: Aroclor 1232
Aroclor 1232

32009: Aroclor 1242
Aroclor 1242

32010: Aroclor 1248
Aroclor 1248

32011: Aroclor 1254
Aroclor 1254

32039: Aroclor 1016/1260
Aroclor 1016/1260

Specifications

Type Multi-component Standard
Quantity 1 mL
ShelfLife 6 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.