Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pesticide Mix 1, SPEX CertiPrep™
SDP

Catalog No. SPXPR1 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SPXPR1 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPXPR1 Supplier SPEX CertiPrep Supplier No. SPXPR1

missing translation for 'validMainframeMsg'

Use these Organic Certified Reference Materials for a wide range of HPLC/GC/LC applications. SPEX CertiPrep™ Pesticide Mix 1 containing 16 compounds in acetronitrile is ideal for pesticide screening, food safety and QuEChERS applications.

  • Pesticide Mix 1 containing 16 compounds in acetronitrile
  • Available and sold in 1mL ampules that are supplied with a pre-labeled transfer/storage vial

Warranty and Services

1 year

Specifications

Chemical Name or Material Trifloxystrobin
Boscali
Fenoxycarb
Piperonyl butoxide
Tebufenpyrad
Packaging Glass Ampule
Assay Percent Range 99.5
99.5
99
95.8
99.5
99.5
99.5
99.5
99.5
99.5
99.5
99.5
99.5
97.6
99.2
99.3
Molecular Formula C20H19F3N2O4
C18H12Cl2N2O
C17H19NO4C19H30O5
C18H24ClN3O
C13H13Cl2N3O3
C9H10ClN5O2
C14H14Cl2N2O
C7H14N2O2S
C7H14N2O3S
C10H9ClN4S
C22H17N3O5
C10H11ClN4
C7H14N2O4S
C11H18N4O2
C18H14BrCl2N5O2
Concentration 100 μg/mL
Quantity 1 mL
Formula Weight 408.37107 g/mol
343.207 g/mol
301.34 g/mol
338.438 g/mol
333.856 g/mol
330.167 g/mol
255.661 g/mol
297.180 g/mol
190.263 g/mol
206.2627 g/mol
252.7233 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.