Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pesticide Mix 10, SPEX CertiPrep™
SDP

Catalog No. SPXPR10 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SPXPR10 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SPXPR10 Supplier SPEX CertiPrep Supplier No. SPXPR10

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use these Organic Certified Reference Materials for a wide range of HPLC/GC/LC applications. SPEX CertiPrep™ Pesticide Mix 10 containing 15 compounds in acetronitrile is ideal for pesticide screening, food safety and QuEChERS applications.

  • Pesticide Mix 9 10 containing 15 compounds in acetronitrile
  • Available and sold in 1mL ampules that are supplied with a pre-labeled transfer/storage vial

Warranty and Services

1 year

Specifications

Chemical Name or Material Aldrin, DDE (p-p')
DDE (p-p')
DDD (o-p)
DDD (p-p')
DDE (o-p)
DDT (o-p')
DDT (p-p')
Packaging Glass Ampule
Assay Percent Range 98.4
99
99.7
99
99.5
99
99
98
97.7
99
99
98
89.4
99.5
99
Molecular Formula C12H8Cl6
C2H2Cl2
C14H10Cl4
C14H10Cl4
C14H8Cl4
C14H9Cl5
C14H9Cl5
C12H8Cl6O
C12H8Cl6O
C12H8Cl6O
C12H8Cl6O
C10H15NO
C10Cl10O
C8H14N4OS
C10Cl12
Concentration 100 μg/mL
Quantity 1 mL
Formula Weight 364.910 g/mol
96.943 g/mol
320.041 g/mol
320.041 g/mol
318.025 g/mol
354.486 g/mol
354.486 g/mol
380.909 g/mol
380.909 g/mol
380.9093 g/mol
380.909 g/mol
165.232 g/mol
490.636 g/mol
214.288 g/mol
545.543 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.