Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pesticide Mix 2, SPEX CertiPrep™
SDP

Catalog No. SPXPR2 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SPXPR2 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPXPR2 Supplier SPEX CertiPrep Supplier No. SPXPR2

missing translation for 'validMainframeMsg'

Use these Organic Certified Reference Materials for a wide range of HPLC/GC/LC applications. SPEX CertiPrep™ Pesticide Mix 2 containing 15 compounds in acetronitrile is ideal for pesticide screening, food safety and QuEChERS applications.

  • Pesticide Mix 2 containing 15 compounds in acetronitrile
  • Available and sold in 1mL ampules that are supplied with a pre-labeled transfer/storage vial

Warranty and Services

1 year

Specifications

Chemical Name or Material Azinphos-methyl
Carbophenothion
Coumaphos
Ethoprophos (ethoprop)
Dimethoate
Dicrotoph
Packaging Glass Ampule
Assay Percent Range 99.5
98.5
99.4
98.8
99.4
99.5
99.2
99.5
98.3
99.5
99.4
99.3
99.5
99.5
99.5
Molecular Formula C10H12N3O3PS2
C11H16ClO2PS3
C14H16ClO5PS
C8H19O2PS2
C5H12NO3PS2
C8H16NO5P
C9H21O2PS3
C12H15N2O3PS
C12H16N3O3PS
C10H15OPS2
C10H19O6PS2
C11H12NO4PS2
C12H15ClNO4PS2
C6H11N2O4PS3
C17H21ClN2O2S
Concentration 100 μg/mL
Quantity 1 mL
Formula Weight 317.324 g/mol
342.865 g/mol
362.766 g/mol
242.339 g/mol
229.257 g/mol
237.190 g/mol
288.431 g/mol
298.298 g/mol
313.313 g/mol
246.329 g/mol
330.358 g/mol
317.321 g/mol
367.809 g/mol
302.331 g/mol
352.879 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.