Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pesticide Mix 3, SPEX CertiPrep™
SDP

Catalog No. SPXPR3 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SPXPR3 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPXPR3 Supplier SPEX CertiPrep Supplier No. SPXPR3

missing translation for 'validMainframeMsg'

Use these Organic Certified Reference Materials for a wide range of HPLC/GC/LC applications. SPEX CertiPrep™ Pesticide Mix 3 containing 15 compounds in acetronitrile is ideal for pesticide screening, food safety and QuEChERS applications.

  • Pesticide Mix 3 containing 15 compounds in acetronitrile
  • Available and sold in 1mL ampules that are supplied with a pre-labeled transfer/storage vial

Warranty and Services

1 year

Specifications

Chemical Name or Material Propargite (omite)
Carbaryl
Myclobutanil (systhane)
Dimethomorph
Etoxazole
Spirodiclo
Packaging Glass Ampule
Assay Percent Range 97.4
99.9
98.6
98.4
98.9
99.5
99.6
99.5
99.5
98.5
99
96
98.9
99
99.4
Molecular Formula C19H26O4S
C12H11NO2
C15H17ClN4
C21H22ClNO4
C21H23F2NO2
C21H24Cl2O4
C8H10ClN5O3S
C9H6F3N3O
C25H28O3
C7H17O2PS3
C2H8NO2PS
C11H15BrClO3PS
C7H14NO5P
C12H17O4PS2
C11H20N3O3PS
Concentration 100 μg/mL
Quantity 1 mL
Formula Weight 350.472 g/mol
201.221 g/mol
288.775 g/mol
387.8567 g/mol
359.410 g/mol
411.319 g/mol
291.715 g/mol
229.159 g/mol
376.488 g/mol
260.377 g/mol
141.001328 g/mol
373.631 g/mol
223.1635 g/mol
320.3647 g/mol
305.334 g/m
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.