Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pesticide Mix 4, SPEX CertiPrep™
SDP

Catalog No. SPXPR4 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SPXPR4 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPXPR4 Supplier SPEX CertiPrep Supplier No. SPXPR4

missing translation for 'validMainframeMsg'

Use these Organic Certified Reference Materials for a wide range of HPLC/GC/LC applications. SPEX CertiPrep™ Pesticide Mix 4 containing 15 compounds in acetronitrile is ideal for pesticide screening, food safety and QuEChERS applications.

  • Pesticide Mix 4 containing 15 compounds in acetronitrile
  • Available and sold in 1mL ampules that are supplied with a pre-labeled transfer/storage vial

Specifications

Chemical Name or Material Epn
Dichlorvos
Edifenphos
Ethion
Fenitrothion
Ethyl parathion
Methyl parathion
Packaging Glass Ampule
Assay Percent Range 99.1
98.8
97.5
99.5
98
99
99.6
99.5
99.5
99.2
99.2
99.5
98
99.5
98
Molecular Formula C14H14NO4PS
C4H7Cl2O4P
C14H15O2PS2
C9H22O4P2S4
C9H12NO5PS
C10H14NO5PS
C8H10NO5PS
C4H10NO3PS
C8H19O2PS3
C10H15O3PS2
C12H21N2O3PS
C9H11Cl3NO3PS
C12H4Cl2F6N4OS
C12H6F2N2O2
C8Cl4N2
Concentration 100 μg/mL
Quantity 1 mL
Formula Weight 323.304 g/mol
220.976 g/mol
310.371 g/mol
384.476 g/mol
277.234 g/mol
291.261 g/mol
263.207 g/mol
183.166 g/mol
274.404 g/mol
278.328 g/mol
304.345 g/mol
350.59 g/mol
437.148 g/mol
248.185 g/mol
265.911 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.