Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pesticide Mix 5, SPEX CertiPrep™
SDP

Catalog No. SPXPR5 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SPXPR5 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPXPR5 Supplier SPEX CertiPrep Supplier No. SPXPR5

missing translation for 'validMainframeMsg'

Use these Organic Certified Reference Materials for a wide range of HPLC/GC/LC applications. SPEX CertiPrep™ Pesticide Mix 5 containing 14 compounds in acetronitrile is ideal for pesticide screening, food safety and QuEChERS applications.

  • Pesticide Mix 5 containing 14 compounds in acetronitrile
  • Available and sold in 1mL ampules that are supplied with a pre-labeled transfer/storage vial

Warranty and Services

1 year

Specifications

Chemical Name or Material Baygon (propoxur)
Metalaxyl
Methomyl
Pymetrozine
Pyraclostrobin
Oxamyl
Paclobutraz
Packaging Glass Ampule
Assay Percent Range 98.2
99.5
99.5
99.9
99.9
99.5
98.6
99.2
99.5
99.5
99.5
99.5
98.4
98.8
Molecular Formula C11H15NO3
C15H21NO4
C5H10N2O2S
C10H11N5O
C19H18ClN3O4
C7H13N3O3S
C15H20ClN3O
C15H16Cl3N3O2
C14H8Cl2N4
C9H10Cl2N2O
C9H10Cl2N2O2
C12H18N2O
C19H21ClN2O
C6H15O4PS2
Concentration 100 μg/mL
Quantity 1 mL
Formula Weight 209.242 g/mol
279.332 g/mol
162.210 g/mol
217.227 g/mol
387.817 g/mol
219.2614 g/mol
293.792 g/mol
376.6654 g/mol
303.146 g/mol
233.094 g/mol
249.094 g/mol
206.284 g/mol
328.836 g/mol
246.2847 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.