Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pesticide Mix 6, SPEX CertiPrep™
SDP

Catalog No. SPXPR6 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SPXPR6 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPXPR6 Supplier SPEX CertiPrep Supplier No. SPXPR6

missing translation for 'validMainframeMsg'

Use these Organic Certified Reference Materials for a wide range of HPLC/GC/LC applications. SPEX CertiPrep™ Pesticide Mix 6 containing 15 compounds in acetronitrile is ideal for pesticide screening, food safety and QuEChERS applications.

  • Pesticide Mix 6 containing 15 compounds in acetronitrile
  • Available and sold in 1mL ampules that are supplied with a pre-labeled transfer/storage vial

Warranty and Services

1 year

Specifications

Chemical Name or Material Fenvalerate (sanmarton)
Pyridaben
Tau-fluvalinate
Quinoxyfen
Alachlor
Prowl (pendimet
Packaging Glass Ampule
Assay Percent Range 99.1
99.5
95
99.7
99.2
99.4
99.5
99.5
98.4
99.4
97.5
99.5
98.9
99
99.5
Molecular Formula C25H22ClNO3
C19H25ClN2OS
C26H22ClF3N2O3
C15H8Cl2FNO
C14H20ClNO2
C13H19N3O4
C18H19NO4
C10H12ClNO2
C17H13ClFN3O
C22H23NO3
C9H7Cl3O3
C10H12N2O3S
C15H22ClNO2
C6Cl5NO2
C9H8Cl3NO2S
Concentration 100 μg/mL
Quantity 1 mL
Formula Weight 419.900 g/mol
364.933 g/mol
502.913 g/mol
308.134 g/mol
269.767 g/mol
281.308 g/mol
313.348 g/mol
213.661 g/mol
329.756 g/mol
349.4229 g/mol
269.509 g/mol
240.279 g/mol
283.794 g/mol
295.335 g/mol
300.589 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.