Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pesticide Mix 7, SPEX CertiPrep™
SDP

Catalog No. SPXPR7 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SPXPR7 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPXPR7 Supplier SPEX CertiPrep Supplier No. SPXPR7

missing translation for 'validMainframeMsg'

Use these Organic Certified Reference Materials for a wide range of HPLC/GC/LC applications. SPEX CertiPrep™ Pesticide Mix 7 containing 8 compounds in acetronitrile is ideal for pesticide screening, food safety and QuEChERS applications.

  • Pesticide Mix 7 containing 8 compounds in acetronitrile
  • Available and sold in 1mL ampules that are supplied with a pre-labeled transfer/storage vial

Warranty and Services

1 year

Specifications

Chemical Name or Material Cypermethrin and an isomer zeta-cypermethrin,
Cyfluthrin (baythroid)
Bifenthrin
Tetramethri
Packaging Glass Ampule
CAS 52645-53-1, 8003-34-7, 10453-86-8, 7696-12-0, 23031-36-9
Assay Percent Range 98.4
99
98
98.2
97.7
98.1
99.4
25
Molecular Formula C22H19Cl2NO3
C22H18Cl2FNO3
C23H22ClF3O2
C19H25NO4
C19H24O3
C21H20Cl2O3
C22H26O3
C22H28O5
Concentration 100 μg/mL
Quantity 1 mL
Formula Weight 416.297 g/mol
434.2876 g/mol
422.8678 g/mol
331.4061 g/mol
300.392 g/mol
391.288 g/mol
338.44 g/mol
372.4547 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.