Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pesticide Mix 8, SPEX CertiPrep™
SDP

Catalog No. SPXPR8 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SPXPR8 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPXPR8 Supplier SPEX CertiPrep Supplier No. SPXPR8

missing translation for 'validMainframeMsg'

Use these Organic Certified Reference Materials for a wide range of HPLC/GC/LC applications. SPEX CertiPrep™ Pesticide Mix 8 containing 15 compounds in acetonitrile is ideal for pesticide screening, food safety and QuEChERS applications.

  • Pesticide Mix 8 containing 15 compounds in acetonitrile
  • Available and sold in 1mL ampules that are supplied with a pre-labeled transfer/storage vial

Warranty and Services

1 year

Specifications

Chemical Name or Material Hexaconazole
Tebuconazole (folicur)
Propiconazole (tilt)
Bifenazate
Spiromesifen
Pine
Packaging Glass Ampule
Assay Percent Range 99.3
99
99.5
99.5
99.5
98
97.5
99.5
99
98.7
99.5
99.6
99.4
98.6
99.5
Molecular Formula C14H17Cl2N3O
C16H22ClN3O
C15H17Cl2N3O2
C17H20N2O3
C23H30O4
C42H69NO10
C48H72O14
C12H17NO2
C11H15NO2S
C9H16ClN5
C11H15NO2
C21H27NO5
C24H27N3O4
C83H132N2O20
C83H132N2O20
Concentration 100 μg/mL
Quantity 1 mL
Formula Weight 314.210 g/mol
307.818 g/mol
342.220 g/mol
300.352 g/mol
370.482 g/mol
747.998 g/mol
873.0769 g/mol
207.269 g/mol
225.307 g/mol
229.710 g/mol
193.242 g/mol
373.443 g/mol
421.489 g/mol
1477.938 g/mol
1477.938 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.