Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pesticide Mix 9, SPEX CertiPrep™
SDP

Catalog No. SPXPR9 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SPXPR9 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPXPR9 Supplier SPEX CertiPrep Supplier No. SPXPR9

missing translation for 'validMainframeMsg'

Use these Organic Certified Reference Materials for a wide range of HPLC/GC/LC applications. SPEX CertiPrep™ Pesticide Mix 9 containing 16 compounds in acetronitrile is ideal for pesticide screening, food safety and QuEChERS applications.

  • Pesticide Mix 9 containing 16 compounds in acetronitrile
  • Available and sold in 1mL ampules that are supplied with a pre-labeled transfer/storage vial

Warranty and Services

1 year

Specifications

Packaging Glass Ampule
Assay Percent Range 97
97
99.5
99.5
96.7
94.2
97
99.5
98.1
98
98.4
98
99.5
99.5
98.6
99.4
Molecular Formula C10H10Cl2O3
C16H12ClNO5
C10H11F3N2O
C14H17Cl2NO2
C4H8Cl3O4P
C13H22NO4PS
C13H22NO5PS
C8H15NOS
C12H15NO4
C12H14N4O4S2
C24H32O4
C12H15NO3
C9H13ClN6
C7H12ClN5
C8H14ClN5
C6H10ClN5
Concentration 100 μg/mL
Quantity 1 mL
Formula Weight 249.091 g/mol
333.723 g/mol
232.202 g/mol
302.196 g/mol
257.437 g/mol
319.357 g/mol
335.356 g/mol
173.276 g/mol
237.252 g/mol
342.394 g/mol
384.509 g/mol
221.252 g/mol
240.693 g/mol
201.657 g/mol
215.683 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.