Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ BL21(DE3) Competent Cells

Catalog No. 694504 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
0.4 mL (2 x 0.2 mL aliquots)
1 mL (5 x 0.2 mL aliquots)
Catalog No. Quantity
69-450-4 1 mL (5 x 0.2 mL aliquots)
69-450-3 0.4 mL (2 x 0.2 mL aliquots)
2 options

Catalog No. 69-450-4

Supplier: MilliporeSigma™ 694504

Only null left
Add to Cart
Add to Cart

Achieve greater efficiency, reproducibility and convenience with Novagen prepared competent cells. BL21(DE3) is a chemically competent E. coli cell suitable for transformation and high-level protein expression using a T7 RNA polymerase-IPTG induction system.

  • Deficient in Ion and ompT proteases, BL21 and its derivatives are ideal for many applications

    T7 expression strain
  • Lysogen of bacteriophage DE3
  • Carries copy of T7 RNA polymerase gene under control of the lacUV5 promoter
  • Suitable for production of protein from target genes cloned in appropriate T7 expression vectors, using IPTG as an inducer
TRUSTED_SUSTAINABILITY

Specifications

Efficiency Guaranteed efficiency: >2 x 107 cfu/μg
Content And Storage Store at ≤-70deg;C, Avoid repeated freeze/thaw
Quantity 1 mL (5 x 0.2 mL aliquots)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.