Learn More
Poltex 6 Pocket PETG Serological Pipette Holder
Description
- Serological Pipette (pipet) Holder.
- Choose from 4, 5, or 6 pocket capacity.
- Choose from powerful, interchangeable magnet mount, 3M VHB (very high bond) tape mount, or countertop placement.
- Add a pipette filler bracket to any version (bracket mounts to either side).
- Magnets and tape mount to either side.
- Break-resistant PETG construction.
- PETG has good alcohol/chemical resistance.
- Shelves are removable for easy cleaning.
Specifications
Specifications
| Material | PETG |
| Positions | 6 Pockets |
| Mounting | Countertop |
| Includes | Pipette Controller Bracket |
| Product Type | Serological Pipet Holder |