Learn More
AquaPhoenix Scientific Petri dish
SCI_ED
Catalog No. S02872 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Material:
Plastic
Catalog No. Material
S02872 Plastic
1 options
Catalog No. S02872 Supplier AquaPhoenix Scientific Supplier No. PD5006
Add to Cart
Add to Cart
Clear plastic Petri dishes for classroom labs and science experiments. Each 90 mm dish is ideal for culturing bacteria, observing mold growth, and supporting hands-on biology activities.
  • Pack of 10 clear plastic Petri dishes
  • Standard 90 mm diameter; height may vary slightly from 14 to 15 mm
  • Ideal for educational use in biology labs or science classrooms
  • Compatible with agar and other common culture media
  • Disposable and lightweight for single-use experiments
  • Suitable for student labs, science fair projects, and STEM instruction
Specifications
Bottom Shape Flat
Color Clear
Gridded No
Material Plastic
Sterility Sterile
Surface Treatment Standard
Type Petri Dishes
Well Count 1
Quantity 10 Pk.
Dimensions (Dia. x H) 90 x 15 mm (or 90 x 14 mm)