Learn More
Carolina Biological supply Petri Dish, approx. 60 x 15 mm
SCI_ED
Catalog No. S000956 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Type:
Petri Dish, 60 x 15 mm
Catalog No. Type
S000956 Petri Dish, 60 x 15 mm
1 options
Catalog No. S000956 Supplier Carolina Biological supply Supplier No. PD5111
Add to Cart
Add to Cart
Dish used for laboratory and experimental applications.
  • Petri dish made of polystyrene designed for scientific and research use
Specifications
Bottom Shape Flat
Color Clear
Gridded No
Material Polystyrene
Sterility Sterile
Surface Treatment Non-treated
Type Petri Dish, 60 x 15 mm
Well Count 1
Height (Metric) 15 mm