Learn More
Research Products International Corp Easyplate Petri Dish Rack, Red, 48 100mm Dish Capacity
SDP
Supplier:  Research Products International Corp 248851
Encompass_Preferred
This rack solves the problem of toppling stacks of petri dishes. Cleverly designed elastic grippers hold plates snuggly in place-even while in transit. Plates store inverted to eliminate buildup of condensation. Available in colors to dress up the lab or help identify plates. Constructed of heavy duty polycarbonate for rugged durability. Autoclavable.
Catalog No. 50-212-619
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.