Learn More
Andwin Scientific PETRI DISH ST 35X10 500/PK
SDP
Supplier:  Andwin Scientific 7067870
Encompass_Preferred
The 35 mm x 10 mm Sterile Petri Dishes are ideal for use in small quantities culture media. This product features three venting ribs into the underside of the lid that prohibits condensation built-up. This product is sold in a pack of 500.
Catalog No. 50-727-390