Learn More
Hach Company Petri Dish with Pad, 9x50 mm, pk/100 (Pall)
Supplier:  Hach Company 110122
Encompass_Preferred
Smaller size standard for water, wastewater, food and beverage analysis using 2.0 mL media. Pre-sterilized, stackable and disposable, these dishes are compatible with 47 mm membrane filters. Double rim ensures tight seal. An area of the lid is frosted for identification. Petri dishes are available with or without pads.
Catalog No. 50-199-5100
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.