Learn More
Andwin Scientific Petri Seal Tape, White, 0.5x108inch
SDP
Supplier:  Andwin Scientific PSEAL108W
Encompass_Preferred
Stretches and conforms to form a tight seal, Temperature resistant, Resists abrasion and puncture, Prevents the entry of contaminant, Designed for microtiter plates, petri dishes, and culture plates.
Catalog No. NC1663546
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.