Learn More
Thermo Scientific™ 3M™ Petrifilm™ E.coli/Coliform Count Plates
SCI_ED
Catalog No. S30729 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
50 Pk.
500 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30729 50 Pk.
S30692 500 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S30729 Supplier Thermo Scientific™ Supplier No. 6404
Add to Cart
Add to Cart
Cost effective, convenient and reliable method for testing equipment, raw materials, food products and manufacturing environmental samples
  • Confirmed E. coli results in 24 to 48 hours
  • Provides accurate, efficient, consistent results for food processors and contract labs
  • Plates are sample-ready, eliminating time-consuming steps of preparing media/agar dishes
  • Interpretation can be done with the 3M™ Petrifilm™ plate reader
  • This plate has been awarded Official Methods of Analysis OMA #991.14.for Food OMA # 998.08 for Poultry, Meat and Sea Food
Specifications
Description For E. coli and coliform
For Use With (Application) Testing equipment, raw materials, food products and manufacturing environmental samples for food processors and contract labs
For Use With (Equipment) 3M™ Petrifilm™ plate reader
Quantity 50 Pk.
Type Environmental test