Learn More
New Pig Petroleum - Based Liquids and Aromatics Drum Gauge
SDP
Catalog No. 17987831 Shop All New Pig Corporation Products
Change view
Click to view available options
Product Type:
Petroleum and Aromatics Drum Fill Gauge
Catalog No. Product Type
17-987-831 Petroleum and Aromatics Drum Fill Gauge
1 options
Catalog No. 17-987-831 Supplier New Pig Supplier No. DRM467
Add to Cart
Add to Cart
Gauge that can be used with flammables has chemical-resistant stainless steel design that's built to last
  • Specifically for petroleum-based liquids: This gauge is designed to be highly compatible with flammables and other petroleum based liquids, ensuring accurate and safe readings.
  • Chemical-resistant construction: Made with durable stainless steel, the gauge is built to withstand a wide range of chemicals and solvents, providing a long service life.
  • Versatile and reliable: It can be used with a variety of liquids, including gasoline, kerosene, diesel fuel, and aromatics, making it a versatile tool for your fluid management needs.
  • Clear and accurate level readings: The gauge's indicator tube provides easy-to-read high and low barrel levels in gallons, liters, and volume percentages.
  • Reduces overflow and spill risks: By providing a clear indication of drum contents, the gauge helps prevent dangerous and costly spills.
  • Built to last: The robust design and materials ensure that this gauge can withstand demanding industrial environments and provide years of reliable service.
Specifications
Product Type Petroleum and Aromatics Drum Fill Gauge
Color Red, Yellow