Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroleum Ether, puriss. p.a., ACS Reagent, Reag. ISO, Solstice

Catalog No. 60046119 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
4 x 5 L
6 x 1 L
195 L
Packaging:
Aluminum Bottle
Glass Bottle
Metal Drum
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-119 1 L Glass Bottle
60-046-125 6 x 1 L Glass Bottle
60-046-122 4 x 2.5 L Glass Bottle
60-046-123 4 x 5 L Aluminum Bottle
60-046-118 195 L Metal Drum
60-046-120 2.5 L Glass Bottle
60-046-124 5 L Aluminum Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 60-046-119 Supplier Solstice Supplier No. 322991L
Add to Cart
Add to Cart

Puriss. p.a., ACS Reagent, Reag. ISO, low boiling point hydrogen treated naphtha, bp≥ 90% 40-60 °C (≥ 90%)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101316-46-5
MDL Number MFCD00081849

Specifications

Appearance Liquid
Density 0.640 to 0.655g/mL at 20°C
Quantity 1 L
Flash Point −30°C (−22°F)
Packaging Glass Bottle
Residue after Evaporation 0 to 0.001%
Titratable Acid Free acid (as CH3COOH): ≤0.002%
UN Number UN1268
Boiling Point 40°C to 60°C
Grade ACS/puriss. p.a.
Chemical Name or Material Petroleum Ether, Reag. ISO
Show More Show Less
Hazard Statement P210-P261-P273-P301 + P310-P331
Precautionary Statement H225-H302-H304-H315-H336-H411
DOTInformation Class 3 - PG 2 - EHS - VP > 110kPa - Petroleum distillates, n.o.s., HI: S
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.