Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroleum Ether, puriss. p.a., Reag. Ph. Eur., Solstice

Catalog No. 60048547 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-048-547 1 L
60-048-548 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-048-547 Supplier Solstice Supplier No. 322471L
Add to Cart
Add to Cart

Chemical Identifiers

CAS 101316-46-5
MDL Number MFCD00081849

Specifications

Density 0.661 to 0.664g/mL at 20°C
Quantity 1 L
Flash Point −26°C (−14.8°F)
UN Number 1268
Grade puriss. p.a.
Chemical Name or Material Petroleum Ether, Reag. Ph. Eur.
Hazard Category Danger
Hazard Statement P201-P210-P301 + P310-P331-P370 + P378-P501
Precautionary Statement H225-H304-H315-H336-H361f-H373-H411
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.