Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroleum ether, for residue analysis, 40-60°C, for trace anal. of polyarom. hydrocarbons

Catalog No. p-216691
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC326790010 1 L Glass Bottle
AC326790025 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC326790010 Supplier Thermo Scientific Chemicals Supplier No. 326790010
Only null left
Add to Cart

CAS: 64742-49-0 | C6H14 | 86.178 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For residue analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64742-49-0
Molecular Formula C6H14
Molecular Weight (g/mol) 86.178
MDL Number MFCD00081849
InChI Key AFABGHUZZDYHJO-UHFFFAOYSA-N
Synonym isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
PubChem CID 7892
IUPAC Name Petroleum ether
SMILES CCCC(C)C

Specifications

Density 0.6500g/mL
Quantity 1 L
Flash Point −45°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3660 to 1.3710
Residue after Evaporation 0.0003% max.
Specific Gravity 0.65
Viscosity 0.45 mm2/s (25°C)
Water 0.005% max. (Coulometric)
Merck Index 14, 5489
Boiling Point 40.0°C to 65.0°C
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with absolute alcohol, benzene,, chloroform, ether, carbon disulfide, carbon, tetrachloride and oils except castor oil
Grade Residue Analysis
Chemical Name or Material Petroleum ether
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement
Highly flammable liquid and vapour.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 232-453-7
RTECSNumber OI6180000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.