Learn More
Specifications
Specifications
| Concentration | 0.2 mg/ml |
| For Use With (Application) | Antibody Competition |
| Formulation | PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin |
| Gene ID (Entrez) | 5192 |
| Quantity | 0.05 mg |
| Storage Requirements | Store at -20°C. Avoid freeze-thaw cycles. |
| Gene Alias | NALD, peroxin 10, Peroxin-10, peroxisomal biogenesis factor 10MGC1998, Peroxisome assembly protein 10, RING finger protein 69, RNF69peroxisome biogenesis factor 10 |
| Gene Symbol | PEX10 |
| Research Category | Zinc Finger |
| Protein | PEX10 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
