Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEX11B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104377
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104377 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104377 Supplier Thermo Scientific Supplier No. PA5104377
Only null left
Add to Cart
Add to Cart

PEX11B Polyclonal Antibody

PEX11B Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen PEX11B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PEX11B
Gene Accession No. O96011, Q9Z210
Gene Alias Peroxin 11 beta; Peroxin 11B; Peroxin-11 beta; Peroxin11B; peroxin-11B; peroxisomal biogenesis factor 11 beta; peroxisomal biogenesis factor 11B; Peroxisomal membrane protein 11B; PEX11 beta; PEX-11 beta; PEX11B; PEX11beta; PEX11-beta; Pex11pbeta; PEX14B; Protein PEX11 homolog beta
Gene Symbols PEX11B
Host Species Rabbit
Immunogen A synthesized peptide derived from human PEX11B(Accession O96011), corresponding to amino acid residues H90-Y140.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18632, 310682, 8799
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.