Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEX11B Recombinant Rabbit Monoclonal Antibody (JE64-51)
GREENER_CHOICE

Catalog No. PIMA544684
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544684 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544684 Supplier Thermo Scientific Supplier No. MA544684
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

PEX11B Recombinant Rabbit Monoclonal Antibody (JE64-51)

PEX11B Recombinant Monoclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen PEX11B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE64-51
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene PEX11B
Gene Accession No. O96011
Gene Alias Peroxin 11 beta; Peroxin 11B; Peroxin-11 beta; Peroxin11B; peroxin-11B; peroxisomal biogenesis factor 11 beta; peroxisomal biogenesis factor 11B; Peroxisomal membrane protein 11B; PEX11 beta; PEX-11 beta; PEX11B; PEX11beta; PEX11-beta; Pex11pbeta; PEX14B; Protein PEX11 homolog beta
Gene Symbols PEX11B
Host Species Rabbit
Immunogen Synthetic peptide within human PEX11B aa 101-150/259.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8799
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.