Learn More
Description
PEX2 Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | PEX2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | PEX2 |
| Gene Accession No. | P28328, P55098 |
| Gene Alias | 35 kDa peroxisomal membrane protein; D3Ertd138e; Paf1; PAF-1; PBD5A; PBD5B; peroxin 2; Peroxin-2; peroxisomal biogenesis factor 2; Peroxisomal membrane protein 3; peroxisomal membrane protein 3, 35 kDa; peroxisomal membrane protein 3, 35kDa; peroxisome assembly factor 1; peroxisome assembly factor-1; peroxisome biogenesis factor 2; Pex2; PMP3; Pmp35; Pxmp3; RING finger protein 72; RNF72; Zellweger syndrome homolog; ZWS3 |
| Gene Symbols | PEX2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
