Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEX26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522033
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522033 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522033 Supplier Invitrogen™ Supplier No. PA522033
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Rhesus Monkey (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene belongs to the peroxin-26 gene family. It is probably required for protein import into peroxisomes. It anchors PEX1 and PEX6 to peroxisome membranes, possibly to form heteromeric AAA ATPase complexes required for the import of proteins into peroxisomes. Defects in this gene are the cause of peroxisome biogenesis disorder complementation group 8. PBD refers to a group of peroxisomal disorders arising from a failure of protein import into the peroxisomal membrane or matrix. The PBD group is comprised of four disorders: Zellweger syndrome, neonatal adrenoleukodystrophy, infantile Refsum disease, and classical rhizomelic chondrodysplasia punctata. Alternatively spliced transcript variants have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEX26
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene PEX26
Gene Accession No. Q7Z412
Gene Alias FLJ20695; PBD7A; PBD7B; Peroxin-26; peroxisomal biogenesis factor 26; peroxisome assembly protein 26; peroxisome biogenesis disorder, complementation group 8; peroxisome biogenesis disorder, complementation group A; peroxisome biogenesis factor 26; PEX26; PEX26M1T; Pex26pM1T
Gene Symbols PEX26
Host Species Rabbit
Immunogen Full length human PEX26 Recombinant protein.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55670
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.