Learn More
Selleck Chemical LLC Pexmetinib (ARRY-614) 5mg 945614-12-0
SDP
Supplier:  Selleck Chemical LLC S77995MG
Encompass_Preferred
Pexmetinib (ARRY-614) is a potent, orally bioavailable, dual p38 MAPK/Tie-2 inhibitor with IC50 of 4 nM/18 nM in a HEK-293 cell line. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8077
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.