Learn More
Medchemexpress LLC Pexopiprant | 932708-14-0 | 99.4% | 456.27 | C21H17Cl2F2NO4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1091865MG
Encompass_Preferred
Pexopiprant is an oral antagonist of the prostaglandin D2 receptor 2 (DP2) used as a research compound for studies such as asthma models. It demonstrates high affinity for DP2 (Ki < 100 nM) and is provided as a high-purity solid with supporting analytical data for use in pharmacology and mechanistic investigations.
  • Oral DP2 antagonist with Ki < 100 nM.
  • High purity suitable for research applications.
  • Soluble in DMSO at 100 mg/mL for convenient dosing.
  • Provided with analytical documentation including COA and spectra.
  • Stable as a powder at -20°C for extended storage.
Catalog No. 50-002-63298
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.